Learn More
Bioss KD-Validated RAD51 Rabbit Reco
SDP
Catalog No. 5000066947
Encompass_Preferred
RAD51 Recombinase Primary Antibody Unconjugated Public Immunogen Range 1-84/339 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66947 Supplier Bioss Supplier No. BSM90032R
RAD51 Recombinase Primary Antibody Unconjugated Public Immunogen Range 1-84/339 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide