Learn More
Bioss KD-Validated RBBP4 Rabbit Reco
SDP
Catalog No. 5000066872
Encompass_Preferred
RB Binding Protein 4 Chromatin Remodeling Factor Primary Antibody Unconjugated Public Immunogen Range 2-46/1066 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66872 Supplier Bioss Supplier No. BSM90465R
RB Binding Protein 4 Chromatin Remodeling Factor Primary Antibody Unconjugated Public Immunogen Range 2-46/1066 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide