Learn More
Bioss KD-Validated Retinoid X recept
SDP
Catalog No. 5000064880
Encompass_Preferred
Retinoid X Receptor Alpha Primary Antibody Unconjugated Public Immunogen Range 1-41/462 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64880 Supplier Bioss Supplier No. BSM90138R
Retinoid X Receptor Alpha Primary Antibody Unconjugated Public Immunogen Range 1-41/462 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide