Learn More
Bioss KD-Validated Rho GTPase activa
SDP
Catalog No. 5000065186
Encompass_Preferred
Rho GTPase Activating Protein 5 Primary Antibody Unconjugated Public Immunogen Range 1150-1250/1502 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65186 Supplier Bioss Supplier No. BSM90167R
Rho GTPase Activating Protein 5 Primary Antibody Unconjugated Public Immunogen Range 1150-1250/1502 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide