Learn More
Bioss KD-Validated RING1 Rabbit Reco
SDP
Catalog No. 5000066167
Encompass_Preferred
Ring Finger Protein 1 Primary Antibody Unconjugated Public Immunogen Range 140-195/406 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66167 Supplier Bioss Supplier No. BSM90345R
Ring Finger Protein 1 Primary Antibody Unconjugated Public Immunogen Range 140-195/406 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide