Learn More
ABclonal Technology KD Validated Sharpin
SDP
Supplier:  ABclonal Technology A2480920UL
Encompass_Preferred
Enables polyubiquitin modification-dependent protein binding activity Involved in protein linear polyubiquitination and regulation of signal transduction Located in cytosol Part of LUBAC complex
Catalog No. 50-000-11282