Learn More
Bioss KD-Validated SMAD5 Rabbit Reco
SDP
Catalog No. 5000066888
Encompass_Preferred
SMAD Family Member 5 Primary Antibody Unconjugated Public Immunogen Range 220-284/465 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66888 Supplier Bioss Supplier No. BSM90140R
SMAD Family Member 5 Primary Antibody Unconjugated Public Immunogen Range 220-284/465 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide