Learn More
Bioss KD-Validated SMYD3 Rabbit Reco
SDP
Catalog No. 5000067020
Encompass_Preferred
SET And MYND Domain Containing 3 Primary Antibody Unconjugated Public Immunogen Range 150-314/314 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67020 Supplier Bioss Supplier No. BSM90504R
SET And MYND Domain Containing 3 Primary Antibody Unconjugated Public Immunogen Range 150-314/314 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide