Learn More
Bioss KD-Validated Sorting Nexin 1 R
SDP
Catalog No. 5000066933
Encompass_Preferred
Sorting Nexin 1 Primary Antibody Unconjugated Public Immunogen Range 1-64/364 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66933 Supplier Bioss Supplier No. BSM90501R
Sorting Nexin 1 Primary Antibody Unconjugated Public Immunogen Range 1-64/364 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide