Learn More
Bioss KD-Validated Src Rabbit Recomb
SDP
Catalog No. 5000066208
Encompass_Preferred
Proto-oncogene tyrosine-protein kinase Src Primary Antibody Unconjugated Public Immunogen Range 1-69/536 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66208 Supplier Bioss Supplier No. BSM90327R
Proto-oncogene tyrosine-protein kinase Src Primary Antibody Unconjugated Public Immunogen Range 1-69/536 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide