Learn More
Bioss KD-Validated STAT1 Rabbit Reco
SDP
Catalog No. 5000066955
Encompass_Preferred
Signal Transducer And Activator Of Transcription 1 Primary Antibody Unconjugated Public Immunogen Range 700-750/750 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66955 Supplier Bioss Supplier No. BSM90182R
Signal Transducer And Activator Of Transcription 1 Primary Antibody Unconjugated Public Immunogen Range 700-750/750 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide