Learn More
Bioss KD-Validated Stat3 Rabbit Reco
SDP
Catalog No. 5000066183
Encompass_Preferred
Signal Transducer And Activator Of Transcription 3 Primary Antibody Unconjugated Public Immunogen Range 670-700/770 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66183 Supplier Bioss Supplier No. BSM90365R
Signal Transducer And Activator Of Transcription 3 Primary Antibody Unconjugated Public Immunogen Range 670-700/770 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide