Learn More
Bioss KD-Validated STAT6 Rabbit Reco
SDP
Catalog No. 5000066533
Encompass_Preferred
Signal Transducer And Activator Of Transcription 6 Primary Antibody Unconjugated Public Immunogen Range 800-847/847 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66533 Supplier Bioss Supplier No. BSM90366R
Signal Transducer And Activator Of Transcription 6 Primary Antibody Unconjugated Public Immunogen Range 800-847/847 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide