Learn More
Bioss KD-Validated Stomatin Like 2 R
SDP
Catalog No. 5000067042
Encompass_Preferred
Stomatin Like 2 Primary Antibody Unconjugated Public Immunogen Range 1500-1531/1531 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67042 Supplier Bioss Supplier No. BSM90485R
Stomatin Like 2 Primary Antibody Unconjugated Public Immunogen Range 1500-1531/1531 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide