Learn More
Bioss KD-Validated STUB1 Rabbit Reco
SDP
Catalog No. 5000065164
Encompass_Preferred
STIP1 Homology And U-Box Containing Protein 1 Primary Antibody Unconjugated Public Immunogen Range 1-51/303 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65164 Supplier Bioss Supplier No. BSM90035R
STIP1 Homology And U-Box Containing Protein 1 Primary Antibody Unconjugated Public Immunogen Range 1-51/303 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide