Learn More
Bioss KD-Validated Superoxide Dismut
SDP
Catalog No. 5000065463
Encompass_Preferred
Superoxide Dismutase 2 Primary Antibody Unconjugated Public Immunogen Range 251-314/428 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65463 Supplier Bioss Supplier No. BSM90503R
Superoxide Dismutase 2 Primary Antibody Unconjugated Public Immunogen Range 251-314/428 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide