Learn More
Bioss KD-Validated Synaptophysin lik
SDP
Catalog No. 5000065418
Encompass_Preferred
Synaptophysin Like 1 Primary Antibody Unconjugated Public Immunogen Range 150-259/259 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65418 Supplier Bioss Supplier No. BSM90203R
Synaptophysin Like 1 Primary Antibody Unconjugated Public Immunogen Range 150-259/259 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide