Learn More
Bioss KD-Validated Syndecan binding
SDP
Catalog No. 5000066831
Encompass_Preferred
Syndecan Binding Protein Primary Antibody Unconjugated Public Immunogen Range 2-79/298 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66831 Supplier Bioss Supplier No. BSM90027R
Syndecan Binding Protein Primary Antibody Unconjugated Public Immunogen Range 2-79/298 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide