Learn More
Bioss KD-Validated TACC3 Rabbit Reco
SDP
Catalog No. 5000065892
Encompass_Preferred
Transforming Acidic Coiled-Coil Containing Protein 3 Primary Antibody Unconjugated Public Immunogen Range 900-1150/1249 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65892 Supplier Bioss Supplier No. BSM90494R
Transforming Acidic Coiled-Coil Containing Protein 3 Primary Antibody Unconjugated Public Immunogen Range 900-1150/1249 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide