Learn More
Bioss KD-Validated TAR DNA binding p
SDP
Catalog No. 5000064882
Encompass_Preferred
TAR DNA Binding Protein Primary Antibody Unconjugated Public Immunogen Range 250-414/414 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64882 Supplier Bioss Supplier No. BSM90143R
TAR DNA Binding Protein Primary Antibody Unconjugated Public Immunogen Range 250-414/414 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide