Learn More
Bioss KD-Validated TEAD1 Rabbit Reco
SDP
Catalog No. 5000064912
Encompass_Preferred
TEA Domain Transcription Factor 1 Primary Antibody Unconjugated Public Immunogen Range 201-248/426 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64912 Supplier Bioss Supplier No. BSM90144R
TEA Domain Transcription Factor 1 Primary Antibody Unconjugated Public Immunogen Range 201-248/426 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide