Learn More
Bioss KD-Validated TGF beta 1 Rabbit
SDP
Catalog No. 5000066652
Encompass_Preferred
Transforming Growth Factor Beta 1 Primary Antibody Unconjugated Public Immunogen Range 50-100/524 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66652 Supplier Bioss Supplier No. BSM90415R
Transforming Growth Factor Beta 1 Primary Antibody Unconjugated Public Immunogen Range 50-100/524 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide