Learn More
Bioss KD-Validated Three Prime Repai
SDP
Catalog No. 5000065464
Encompass_Preferred
Three Prime Repair Exonuclease 1 Primary Antibody Unconjugated Public Immunogen Range 770-835/858 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65464 Supplier Bioss Supplier No. BSM90505R
Three Prime Repair Exonuclease 1 Primary Antibody Unconjugated Public Immunogen Range 770-835/858 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide