Learn More
Bioss KD-Validated Topoisomerase II
SDP
Catalog No. 5000066799
Encompass_Preferred
DNA Topoisomerase II Alpha Primary Antibody Unconjugated Public Immunogen Range 362-400/400 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66799 Supplier Bioss Supplier No. BSM90486R
DNA Topoisomerase II Alpha Primary Antibody Unconjugated Public Immunogen Range 362-400/400 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide