Learn More
Bioss KD-Validated Transcription elo
SDP
Catalog No. 5000066536
Encompass_Preferred
Transcription Elongation Factor A1 Primary Antibody Unconjugated Public Immunogen Range 350-427/427 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66536 Supplier Bioss Supplier No. BSM90396R
Transcription Elongation Factor A1 Primary Antibody Unconjugated Public Immunogen Range 350-427/427 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide