Learn More
ABclonal Technology KD Validated TRIM25 Rabbit mAb
SDP
Catalog No. 5000010759
Encompass_Preferred
The protein encoded by this gene is a member of the tripartite motif (TRIM) family The TRIM motif includes three zinc-binding domains a RING a B-box type 1 and a B-box type 2 and a coiled-coil region The protein is an RNA binding protein functions as a ubiquitin E3 ligase and is involved in multiple cellular processes including regulation of antiviral innate immunity
Catalog No. 50-000-10759 Supplier ABclonal Technology Supplier No. A25846100UL
The protein encoded by this gene is a member of the tripartite motif (TRIM) family The TRIM motif includes three zinc-binding domains a RING a B-box type 1 and a B-box type 2 and a coiled-coil region The protein is an RNA binding protein functions as a ubiquitin E3 ligase and is involved in multiple cellular processes including regulation of antiviral innate immunity