Learn More
Bioss KD-Validated Tripartite motif
SDP
Catalog No. 5000066999
Encompass_Preferred
Tripartite Motif Containing 24 Primary Antibody Unconjugated Public Immunogen Range 150-200/796 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66999 Supplier Bioss Supplier No. BSM90409R
Tripartite Motif Containing 24 Primary Antibody Unconjugated Public Immunogen Range 150-200/796 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide