Learn More
Bioss KD-Validated TRK fused gene Ra
SDP
Catalog No. 5000066800
Encompass_Preferred
Trafficking From ER To Golgi Regulator Primary Antibody Unconjugated Public Immunogen Range 350-494/494 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66800 Supplier Bioss Supplier No. BSM90487R
Trafficking From ER To Golgi Regulator Primary Antibody Unconjugated Public Immunogen Range 350-494/494 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide