Learn More
Bioss KD-Validated Trophoblast Glyco
SDP
Catalog No. 5000067025
Encompass_Preferred
Trophoblast Glycoprotein Primary Antibody Unconjugated Public Immunogen Range 230-280/366 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67025 Supplier Bioss Supplier No. BSM90508R
Trophoblast Glycoprotein Primary Antibody Unconjugated Public Immunogen Range 230-280/366 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide