Learn More
Bioss KD-Validated Tubulin Beta 3 Cl
SDP
Catalog No. 5000067021
Encompass_Preferred
Tubulin Beta 3 Class III Primary Antibody Unconjugated Public Immunogen Range 15-50/1302 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67021 Supplier Bioss Supplier No. BSM90491R
Tubulin Beta 3 Class III Primary Antibody Unconjugated Public Immunogen Range 15-50/1302 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide