Learn More
Bioss KD-Validated Ubiquitin specifi
SDP
Supplier:  Bioss BSM90184R
Encompass_Preferred
Ubiquitin Specific Peptidase 13 Primary Antibody Unconjugated Public Immunogen Range 750-800/863 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66891