Learn More
Bioss KD-Validated Ubiquitin Specifi
SDP
Catalog No. 5000066824
Encompass_Preferred
Ubiquitin Specific Peptidase 14 Primary Antibody Unconjugated Public Immunogen Range 300-375/453 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66824 Supplier Bioss Supplier No. BSM90488R
Ubiquitin Specific Peptidase 14 Primary Antibody Unconjugated Public Immunogen Range 300-375/453 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide