Learn More
Bioss KD-Validated Ubiquitin Specifi
SDP
Supplier:  Bioss BSM90506R
Encompass_Preferred
Ubiquitin Specific Peptidase 5 Primary Antibody Unconjugated Public Immunogen Range 50-100/704 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66828