Learn More
Bioss KD-Validated Ubiquitin specifi
SDP
Catalog No. 5000064871
Encompass_Preferred
Ubiquitin Specific Peptidase 7 Primary Antibody Unconjugated Public Immunogen Range 100-150/1102 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64871 Supplier Bioss Supplier No. BSM90030R
Ubiquitin Specific Peptidase 7 Primary Antibody Unconjugated Public Immunogen Range 100-150/1102 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide