Learn More
Bioss KD-Validated Vasodilator stimu
SDP
Catalog No. 5000064975
Encompass_Preferred
Vasodilator Stimulated Phosphoprotein Primary Antibody Unconjugated Public Immunogen Range 44-94/380 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64975 Supplier Bioss Supplier No. BSM90206R
Vasodilator Stimulated Phosphoprotein Primary Antibody Unconjugated Public Immunogen Range 44-94/380 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide