Learn More
ABclonal Technology KD Validated XPNPEP3
SDP
Supplier:  ABclonal Technology A2695020UL
Encompass_Preferred
The protein encoded by this gene belongs to the family of X-pro-aminopeptidases that utilize a metal cofactor and remove the N-terminal amino acid from peptides with a proline residue in the penultimate position This protein has been shown to localize to the mitochondria of renal cells and have a role in ciliary function Mutations in this gene are associated with nephronophthisis-like nephropathy-1 Alternatively spliced transcript variants encoding different isoforms have been noted for this gene however expression of some of these isoforms in vivo is not known
Catalog No. 50-000-12466