Learn More
Bioss KD-Validated ZPR1 Zinc Finger
SDP
Catalog No. 5000065894
Encompass_Preferred
ZPR1 Zinc Finger Primary Antibody Unconjugated Public Immunogen Range 1000-1093/1093 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65894 Supplier Bioss Supplier No. BSM90511R
Add to Cart
Add to Cart
ZPR1 Zinc Finger Primary Antibody Unconjugated Public Immunogen Range 1000-1093/1093 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide