Learn More
Selleck Chemical LLC KD025 (SLx-2119) 5mg 911417-87-3
SDP
Catalog No. NC1332801
Encompass_Preferred
KD025 (SLx-2119) is an orally available, and selective ROCK2 inhibitor with IC50 and Ki of 60 nM and 41 nM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1332801 Supplier Selleck Chemical LLC Supplier No. S79365MG
KD025 (SLx-2119) is an orally available, and selective ROCK2 inhibitor with IC50 and Ki of 60 nM and 41 nM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.