Learn More
Enzo Life Sciences KDEL monoclonal antibody (10C3) (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPA827J
Encompass_Preferred
Clone: 10C3. Host: Mouse. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 649-654 (S649EKDEL654) of rat Grp78. UniProt ID: P06761. Source: Purified from ascites. Species reactivity: Yeast, Plant, Mammalian, Insect, Avian. Specificity: Recognizes proteins containing the KDEL sequence. Applications: ELISA, Flow Cytometry, ICC, IF, IHC (PS), IP, WB Electron microscopy. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7527