Learn More
Enzo Life Sciences KDEL monoclonal antibody (10C3) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6790200
Encompass_Preferred
Clone: 10C3. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 649-654 (S649EKDEL654) of rat Grp78. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Yeast, Plant, Mammalian, Insect, Avian. Specificity: Recognizes proteins containing the KDEL sequence. Applications: ELISA, Flow Cytometry, ICC, IF, IHC (PS), IP, WB Electron microscopy. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3209