Learn More
Enzo Life Sciences KDEL monoclonal antibody (10C3) (DyLight 488 conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA827488F
Encompass_Preferred
Clone: 10C3. Host: Mouse. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 649-654 (S649EKDEL654) of rat Grp78. Source: Purified from ascites. Species reactivity: Yeast, Plant, Mammalian, Insect, Avian. Specificity: Recognizes proteins containing the KDEL sequence. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-200-7526