Learn More
Enzo Life Sciences KDEL receptor monoclonal antibody (KR-10) (50µl)
SDP
Supplier:  Enzo Life Sciences ADIVAAPT048D
Encompass_Preferred
Clone: KR-10. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of bovine KDEL Receptor. UniProt ID: P33946 (KDELR1), Q2KJ37 (KDELR2). Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Recommended Dilutions: Western Blot (1:250, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ascites. Formulation: Liquid. Mouse ascites fluid containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7681