Learn More
Aviva Systems Biology Corp KDELR3 Antibody - middle region (ARP46794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KDELR3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KDELR3. Alternate names: ERD2L3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59402