Learn More
TARGETMOL CHEMICALS INC KDM2B-IN-2 5MG
SDP
Catalog No. 502580055
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KDM2B-IN-2 is a potent histone demethylase (kdm2b) inhibitor. purity: 97%
Catalog No. 50-258-0055 Supplier TARGETMOL CHEMICALS INC Supplier No. T393895MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KDM2B-IN-2 is a potent histone demethylase (kdm2b) inhibitor. purity: 97%