Learn More
Medchemexpress LLC KDM2B-IN-4 | 1966933-85-6 | >99.6% | 376.49 | 50 MG
SDP
Catalog No. 5000348927
Encompass_Preferred
KDM2B-IN-4 is a potent histone demethylase KDM2B inhibitor with an IC50 of 1.12 nM. This compound is suitable for research into hyperproliferative diseases, including various types of cancers.
  • Potent histone demethylase KDM2B inhibitor
  • IC50 of 1.12 nM
  • Useful for hyperproliferative disease research
  • Applicable in cancer research
Catalog No. 50-003-48927 Supplier Medchemexpress LLC Supplier No. HY13960150MG
KDM2B-IN-4 is a potent histone demethylase KDM2B inhibitor with an IC50 of 1.12 nM. This compound is suitable for research into hyperproliferative diseases, including various types of cancers.
  • Potent histone demethylase KDM2B inhibitor
  • IC50 of 1.12 nM
  • Useful for hyperproliferative disease research
  • Applicable in cancer research