Learn More
ABclonal Technology KDM4A Rabbit mAb
SDP
Catalog No. 5000007588
Encompass_Preferred
This gene is a member of the Jumonji domain 2 (JMJD2) family and encodes a protein containing a JmjN domain a JmjC domain a JD2H domain two TUDOR domains and two PHD-type zinc fingers This nuclear protein functions as a trimethylation-specific demethylase converting specific trimethylated histone residues to the dimethylated form and as a transcriptional repressor
Catalog No. 50-000-07588 Supplier ABclonal Technology Supplier No. A2206020UL
This gene is a member of the Jumonji domain 2 (JMJD2) family and encodes a protein containing a JmjN domain a JmjC domain a JD2H domain two TUDOR domains and two PHD-type zinc fingers This nuclear protein functions as a trimethylation-specific demethylase converting specific trimethylated histone residues to the dimethylated form and as a transcriptional repressor