Learn More
Sino Biological Recombinant Mouse KDM1B Protein (Full Length, GST Tag)
Catalog No. 502574515
Encompass_Preferred
Recombinant full length mouse KDM1B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4515 Supplier Sino Biological Supplier No. K42130BG100
Recombinant full length mouse KDM1B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.