Learn More
TARGETMOL CHEMICALS INC KDM4D-IN-1 5MG
SDP
Catalog No. 502579925
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KDM4D-IN-1 is a KDM4D inhibitor (IC50 0.41±0.03 uM). purity: 99%
Catalog No. 50-257-9925 Supplier TARGETMOL CHEMICALS INC Supplier No. T42145MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KDM4D-IN-1 is a KDM4D inhibitor (IC50 0.41±0.03 uM). purity: 99%