Learn More
Medchemexpress LLC HY-101928 5mg Medchemexpress, KDM4D-IN-1 CAS:2098902-68-0 Purity:>98%
SDP
Catalog No. 501967919
Encompass_Preferred
Medchemexpress, HY-101928 5mg KDM4D-IN-1 CAS:2098902-68-0 KDM4D-IN-1 is a new histone lysine demethylase 4D (KDM4D) inhibitor with an IC50 value of 0.41±0.03 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7919 Supplier Medchemexpress LLC Supplier No. HY1019285MG
Medchemexpress, HY-101928 5mg KDM4D-IN-1 CAS:2098902-68-0 KDM4D-IN-1 is a new histone lysine demethylase 4D (KDM4D) inhibitor with an IC50 value of 0.41±0.03 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.