Learn More
Medchemexpress LLC HY-120400 25mg Medchemexpress, KDM5-C70 CAS:1596348-32-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12040025MG
Encompass_Preferred
Medchemexpress, HY-120400 25mg KDM5-C70 CAS:1596348-32-1 KDM5-C70 is an ethyl ester derivative of KDM5-C49 and a potent, cell-permeable and pan-KDM5 histone demethylase inhibitor. KDM5-C70 has an antiproliferative effect in myeloma cells, leading to genome-wide elevation of H3K4me3 levels[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8842